Learn More
Cayman Chemical Dapoxetine (hydrochloride), Purity: ≥98%, CAS: 129938-20-1, 50 mg
SDP
Catalog No. 502894185
Encompass_Preferred
An analytical reference standard categorized as an SSRI; intended for research and forensic applications
Catalog No. 50-289-4185 Supplier Cayman Chemical Supplier No. 900248850 MG
An analytical reference standard categorized as an SSRI; intended for research and forensic applications