Learn More
Aviva Systems Biology Corp DAPP1 Antibody - middle region (ARP55041_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55041_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DAPP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DAPP1. Alternate names: BAM32, DKFZp667E0716ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-992