Learn More
Medchemexpress LLC HY-17608 10mg Medchemexpress, Daprodustat CAS:960539-70-2 Purity:>98%
SDP
Catalog No. 502029249
Encompass_Preferred
Medchemexpress, HY-17608 10mg Daprodustat CAS:960539-70-2 Daprodustat (GSK1278863) is an orally active hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor being developed for the treatment of anemia associated with chronic kidney disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9249 Supplier Medchemexpress LLC Supplier No. HY1760810MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-17608 10mg Daprodustat CAS:960539-70-2 Daprodustat (GSK1278863) is an orally active hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor being developed for the treatment of anemia associated with chronic kidney disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.