Learn More
Medchemexpress LLC HY-17608 50mg , Daprodustat CAS:960539-70-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1760850MG
Encompass_Preferred
HY-17608 50mg Daprodustat CAS: 960539-70-2 Daprodustat (GSK1278863) is an orally active hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor being developed for the treatment of anemia associated with chronic kidney disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0211