Learn More
Apexbio Technology LLC Daprodustat(GSK1278863), 10mg Cas No: 960539-70-2 MFCD: MFCD29924726
SDP
Supplier:  Apexbio Technology LLC B610610
Encompass_Preferred
Daprodustat (GSK1278863) is a HIF-prolyl hydroxylase inhibitor.Hypoxia stimulates erythropoietin (EPO) release through accumulation of hypoxia-inducible factor 1 (HIF-1), resulting in an increase of erythrocyte production by the bone marrow. There are other sizes available. Please inqury us for quote.
Catalog No. NC1448343