Learn More
Apexbio Technology LLC Dapsone 80-08-0 1g
SDP
Catalog No. 502240669
Encompass_Preferred
Dapsone (80-08-0) is a small-molecule inhibitor targeting dihydropteroate synthase (DHPS) It is designed to interfere with folate synthesis pathways thereby impeding bacterial nucleic acid production Dapsone exerts its biological activity primarily through DHPS inhibition In in vitro studies dapsone demonstrates inhibitory activity with IC50 values typically ranging between 0 1 1 M dependent on organism strain and experimental conditions Based on these pharmacological properties dapsone holds research potential in leprosy drug development adjunctive antimicrobial therapy investigation antimicrobial resistance profiling and in studies targeting protozoal parasites such as Plasmodium falciparum
This item is not returnable. View return policy
Catalog No. 50-224-0669 Supplier Apexbio Technology LLC Supplier No. B34481000
Dapsone (80-08-0) is a small-molecule inhibitor targeting dihydropteroate synthase (DHPS) It is designed to interfere with folate synthesis pathways thereby impeding bacterial nucleic acid production Dapsone exerts its biological activity primarily through DHPS inhibition In in vitro studies dapsone demonstrates inhibitory activity with IC50 values typically ranging between 0 1 1 M dependent on organism strain and experimental conditions Based on these pharmacological properties dapsone holds research potential in leprosy drug development adjunctive antimicrobial therapy investigation antimicrobial resistance profiling and in studies targeting protozoal parasites such as Plasmodium falciparum