Learn More
Medchemexpress LLC Dapsone (Standard) | 80-08-0 | 99.8% | 250 MG
SDP
Catalog No. 5000406570
Encompass_Preferred
Dapsone (Standard) is the analytical standard for Dapsone, an orally active, blood-brain penetrant sulfonamide antibiotic with bacteriostatic, antimycobacterial, and antiprotozoal activities. It inhibits folate synthesis in M. leprae cell extracts and is used in dermatologic disorder research, including leprosy, dermatitis herpetiformis, and acne vulgaris. This product is intended for research and analytical applications.
  • Analytical standard for Dapsone
  • Exhibits bacteriostatic, antimycobacterial, and antiprotozoal activities
  • Inhibits folate synthesis in M. leprae
  • Used in dermatologic disorder research
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Commonly used in HPLC, GC, and MS applications
Catalog No. 50-004-06570 Supplier Medchemexpress LLC Supplier No. 250MGHYB0688R
Add to Cart
Add to Cart
Dapsone (Standard) is the analytical standard for Dapsone, an orally active, blood-brain penetrant sulfonamide antibiotic with bacteriostatic, antimycobacterial, and antiprotozoal activities. It inhibits folate synthesis in M. leprae cell extracts and is used in dermatologic disorder research, including leprosy, dermatitis herpetiformis, and acne vulgaris. This product is intended for research and analytical applications.
  • Analytical standard for Dapsone
  • Exhibits bacteriostatic, antimycobacterial, and antiprotozoal activities
  • Inhibits folate synthesis in M. leprae
  • Used in dermatologic disorder research
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Commonly used in HPLC, GC, and MS applications