Learn More
Cayman Chemical DAPT, 208255-80-5, 5 mg
SDP
Supplier:  Cayman Chemical 131975
Encompass_Preferred
Molecular Formula C23H26F2N2O4, Purity ≥95%, Formula Weight 432.5, Synonyms: N-[2S-(3,5-difluorophenyl)acetyl]-L-alanyl-2-phenyl-1,1-dimethylethyl ester-glycine, GSI-IX; An inhibitor of -secretase, blocking the production of total A in human primary neuronal cultures with an IC50 value of 115 nM and A42 with an IC50 value of 200 nM; reduces brain levels of A in vivo when given orally; indirectly inhibits Notch, affecting cell signaling and cell differentiation.
Catalog No. NC0774775
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.