Learn More
Medchemexpress LLC HY-13027 50mg , DAPT GSI-IX CAS:208255-80-5 Purity:98%
SDP
Catalog No. 501493625
Encompass_Preferred
Medchemexpress, HY-13027 50mg DAPT GSI-IX CAS:208255-80-5 Formula:C23H26F2N2O4 IC50: 20 nM (Aβ, in HEK 293 cells) Purity:98% DAPT is a γ-secretase inhibitor, reduces the total beta-amyloid peptide ( Aβ ) production with IC 50 of 20 nM in HEK 293 cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3625 Supplier Medchemexpress LLC Supplier No. HY1302750MG
Medchemexpress, HY-13027 50mg DAPT GSI-IX CAS:208255-80-5 Formula:C23H26F2N2O4 IC50: 20 nM (Aβ, in HEK 293 cells) Purity:98% DAPT is a γ-secretase inhibitor, reduces the total beta-amyloid peptide ( Aβ ) production with IC 50 of 20 nM in HEK 293 cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.