Learn More
Enzo Life Sciences DAPT, (5mg), CAS Number: 208255-80-5
SDP
Supplier:  Enzo Life Sciences ALX270416M005
Encompass_Preferred
γ-Secretase inhibitor. Alternative Name: N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine tert-butyl ester, gamma-Secretase inhibitor IX. Formula: C23H26F2N2O4. MW: 432.5. CAS Number: 208255-80-5. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in 100% ethanol, DMSO (20mg/ml) or dichloromethane. Use/Stability: Stock solutions are stable for up to 3 months when stored at -20°C. Long Term Storage: -20°C
Catalog No. 50-103-1740