Learn More
Medchemexpress LLC HY-P1034 1mg Medchemexpress, DAPTA CAS:106362-34-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP10341MG
Encompass_Preferred
Medchemexpress, HY-P1034 1mg DAPTA CAS:106362-34-9 DAPTA is a synthetic peptide, functions as a viral entry inhibitor by targeting selectively CCR5 , and shows potent anti-HIV activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4143