Learn More
Cayman Chemical DAPTA (acetate), Purity: ≥98%, 50 mg
SDP
Catalog No. 502948540
Encompass_Preferred
A CCR5 receptor antagonist; inhibits CD4-dependent binding of HIV-1Ba-L and HIV-1CM235 gp120 envelope proteins to CCR5 (IC50s = 0.06 and 0.32 nM, respectively); reduces marble burying and repetitive behavior, as well as decreases brain levels of IL-6, IL-9, and IL-17A, in the BTBR T+ Itpr3tf/J mouse model of ASD at 0.1 mg/kg; reduces LPS-induced microglial activation and astrocyte hypertrophy in the dentate gyrus in a rat model of chronic neuroinflammation
Catalog No. 50-294-8540 Supplier Cayman Chemical Supplier No. 3333950 MG
A CCR5 receptor antagonist; inhibits CD4-dependent binding of HIV-1Ba-L and HIV-1CM235 gp120 envelope proteins to CCR5 (IC50s = 0.06 and 0.32 nM, respectively); reduces marble burying and repetitive behavior, as well as decreases brain levels of IL-6, IL-9, and IL-17A, in the BTBR T+ Itpr3tf/J mouse model of ASD at 0.1 mg/kg; reduces LPS-induced microglial activation and astrocyte hypertrophy in the dentate gyrus in a rat model of chronic neuroinflammation