Learn More
TARGETMOL CHEMICALS INC Daptomycin 100MG
SDP
Catalog No. 502876077
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Daptomycin (LY146032) is a cyclic lipopeptide antibiotic that inhibits GRAM-POSITIVE BACTERIA. Purity 99.61%
Catalog No. 50-287-6077 Supplier TARGETMOL CHEMICALS INC Supplier No. T1665100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Daptomycin (LY146032) is a cyclic lipopeptide antibiotic that inhibits GRAM-POSITIVE BACTERIA. Purity 99.61%