Learn More
Enzo Life Sciences Daptomycin (100mg). CAS: 103060-53-3
SDP
Supplier:  Enzo Life Sciences BMLA2010100
Encompass_Preferred
Novel lipopeptide antibiotic active against Gram positive bacteria. Alternative name: Cubicin, LY146032. Purity: ≥93% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: +4°C.
Catalog No. 50-201-0231