Learn More
Medchemexpress LLC Daptomycin | 103060-53-3 | 99.7% | C72H101N17O26 | 25 MG
SDP
Catalog No. 5000357887
Encompass_Preferred
Daptomycin is a lipopeptide antibiotic with rapid in vitro bactericidal activity against gram-positive organisms.
  • Rapid in vitro bactericidal activity against gram-positive organisms.
  • Excellent in-vitro inhibitory and bactericidal activity against nafcillin-susceptible and resistant staphylococci (MIC90 less than or equal to 0.5 mg/L) and against enterococci (MIC90 less than or equal to 2.0 mg/L).
  • More active than vancomycin against the majority of isolated tested.
  • Most active agent in vitro against enterococci, and most active against nafcillin-resistant staphylococci (with the exception of trimethoprim-sulphamethoxazole).
Catalog No. 50-003-57887 Supplier Medchemexpress LLC Supplier No. 25MGHYB0108
Daptomycin is a lipopeptide antibiotic with rapid in vitro bactericidal activity against gram-positive organisms.
  • Rapid in vitro bactericidal activity against gram-positive organisms.
  • Excellent in-vitro inhibitory and bactericidal activity against nafcillin-susceptible and resistant staphylococci (MIC90 less than or equal to 0.5 mg/L) and against enterococci (MIC90 less than or equal to 2.0 mg/L).
  • More active than vancomycin against the majority of isolated tested.
  • Most active agent in vitro against enterococci, and most active against nafcillin-resistant staphylococci (with the exception of trimethoprim-sulphamethoxazole).