Learn More
Medchemexpress LLC Daptomycin | 103060-53-3 | 99.7% | C72H101N17O26 | 5 MG
SDP
Catalog No. 5000432443
Encompass_Preferred
Daptomycin is a lipopeptide antibiotic with rapid in vitro bactericidal activity against gram-positive organisms.
  • Exhibits excellent inhibitory and bactericidal activity against nafcillin-susceptible and resistant staphylococci and enterococci.
  • More active than vancomycin against the majority of isolated tested.
  • Shows high activity against nafcillin-resistant staphylococci.
  • Inhibits the formation of nucleotide-linked intermediates.
Catalog No. 50-004-32443 Supplier Medchemexpress LLC Supplier No. HYB01085MG
Daptomycin is a lipopeptide antibiotic with rapid in vitro bactericidal activity against gram-positive organisms.
  • Exhibits excellent inhibitory and bactericidal activity against nafcillin-susceptible and resistant staphylococci and enterococci.
  • More active than vancomycin against the majority of isolated tested.
  • Shows high activity against nafcillin-resistant staphylococci.
  • Inhibits the formation of nucleotide-linked intermediates.