Learn More
Medchemexpress LLC Daptomycin | 103060-53-3 | MFCD08282794 | 1620.67 | C72H101N17O26 | 10 MG
SDP
Catalog No. 5000297385
Encompass_Preferred
Daptomycin is a lipopeptide antibiotic with rapid in vitro bactericidal activity against gram-positive organisms. Supplied as a research-grade powder for laboratory use, it can be formulated for in vitro and in vivo studies according to manufacturer solubility guidance. Store sealed and protected from moisture; powder stability is specified at -80°C (2 years) and -20°C (1 year).
  • Lipopeptide antibiotic active against gram-positive organisms.
  • Suitable for in vitro and in vivo research applications.
  • Supplied as a powder for flexible formulation and dosing.
  • Soluble in water and DMSO; ultrasonic assistance may be required.
  • Sealed storage recommended; powder stable at -80°C and -20°C per datasheet.
  • Available in small research pack sizes, including 10 mg.
Catalog No. 50-002-97385 Supplier Medchemexpress LLC Supplier No. HYB010810MG
Daptomycin is a lipopeptide antibiotic with rapid in vitro bactericidal activity against gram-positive organisms. Supplied as a research-grade powder for laboratory use, it can be formulated for in vitro and in vivo studies according to manufacturer solubility guidance. Store sealed and protected from moisture; powder stability is specified at -80°C (2 years) and -20°C (1 year).
  • Lipopeptide antibiotic active against gram-positive organisms.
  • Suitable for in vitro and in vivo research applications.
  • Supplied as a powder for flexible formulation and dosing.
  • Soluble in water and DMSO; ultrasonic assistance may be required.
  • Sealed storage recommended; powder stable at -80°C and -20°C per datasheet.
  • Available in small research pack sizes, including 10 mg.