Learn More
AdipoGen Daptomycin
SDP
Catalog No. 502562481
Encompass_Preferred
Chemical. CAS 103060-53-3. Formula C72H101N17O26. MW 1620.7. Daptomycin, a naturally occurring cyclic lipopeptide, is a calcium-dependent antibiotic. It exhibits potent bacteriocidal activity against most Gram-positive bacteria in vitro and in vivo, including antibiotic-resistant strains such as MRSA and VRE. Daptomycin disrupts the bacterial plasma membrane function, which results in membrane depolarization consequently leading to inhibition of protein, DNA and RNA synthesis, essential for cell division and cell wall synthesis.
Catalog No. 50-256-2481 Supplier AdipoGen Supplier No. AGCN20542M100
Chemical. CAS 103060-53-3. Formula C72H101N17O26. MW 1620.7. Daptomycin, a naturally occurring cyclic lipopeptide, is a calcium-dependent antibiotic. It exhibits potent bacteriocidal activity against most Gram-positive bacteria in vitro and in vivo, including antibiotic-resistant strains such as MRSA and VRE. Daptomycin disrupts the bacterial plasma membrane function, which results in membrane depolarization consequently leading to inhibition of protein, DNA and RNA synthesis, essential for cell division and cell wall synthesis.