Learn More
Selleck Chemical LLC Daptomycin S1373-10mM/1mL
SDP
Catalog No. 503108721
Encompass_Preferred
Daptomycin is a novel antibiotic with rapid in vitro bactericidal activity against gram-positive organisms
Catalog No. 50-310-8721 Supplier Selleck Chemical LLC Supplier No. S137310MM/1ML
Daptomycin is a novel antibiotic with rapid in vitro bactericidal activity against gram-positive organisms