Learn More
Abcam Daptomycin, Lipopeptide antibiotic, 10MG
Catalog No. 502706804
Encompass_Preferred
MW 1620.7 Da. Lipopeptide antibiotic. Gram-positive bacterial membrane integrity antagonist. Does not interact with p450 cytochromes. Concentration-dependent antibacterial activity in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6804 Supplier Abcam Supplier No. AB14120410MG
MW 1620.7 Da. Lipopeptide antibiotic. Gram-positive bacterial membrane integrity antagonist. Does not interact with p450 cytochromes. Concentration-dependent antibacterial activity in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement