Learn More
AdipoGen DAR-1
SDP
Supplier:  AdipoGen CDXD0101M005
Encompass_Preferred
Chemical. CAS 261351-43-3. Formula C28H32N4O3. MW 472.58. Synthetic. Sensitive NO probe, LOD of 10 nM, shows higher photostability than the classical fluorescein derivative DAF. Spectral data lambdaex 566nm lambdaem 586nm in 0.1 M phosphate pH 7.4.
Catalog No. 50-256-1238