Learn More
Abcam DAR-1, Fluorescent NO probe, 1MG
Catalog No. 502705241
Encompass_Preferred
MW 472.58 g/mol, Purity >97%. Sensitive fluorescent NO probe. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5241 Supplier Abcam Supplier No. AB1453881MG
MW 472.58 g/mol, Purity >97%. Sensitive fluorescent NO probe. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement