Learn More
AdipoGen DAR-1T Solution
SDP
Supplier:  AdipoGen CDXD0513M001
Encompass_Preferred
Chemical. CAS 261351-46-6. Formula C28H29N5O3. MW 483.57. Synthetic Reference compound for the fluorescent nitric oxide NO probe DAR-1.
Catalog No. 50-256-1060