Learn More
AdipoGen DAR-2
SDP
Supplier:  AdipoGen CDXD0102M025
Encompass_Preferred
Chemical. CAS 261351-45-5. Formula C28H32N4O3. MW 472.58. Synthetic. Sensitive NO probe, LOD of 10 nM, shows higher photostability than the classical fluorescein derivative DAF. Spectral data lambdaex 562nm lambdaem 579nm in 0.1 M phosphate pH 7.4.
Catalog No. 50-256-1234