Learn More
AdipoGen DAR-2T
SDP
Supplier:  AdipoGen CDXD0214M001
Encompass_Preferred
Chemical. CAS 261351-48-8. Formula C28H29N5O3. MW 483.57. Synthetic. Reference compound for the fluorescent nitric oxide NO probe DAR-2.
Catalog No. 50-256-1180