Learn More
AdipoGen DAR-4 Solution
SDP
Catalog No. 502561052
Encompass_Preferred
Chemical. CAS 339527-78-5. Formula C24H24N4O3. MW 416.48. Synthetic Photo-stable fluorescent probe for the detection of nitric oxide NO.
Catalog No. 50-256-1052 Supplier AdipoGen Supplier No. CDXD0576M001
Chemical. CAS 339527-78-5. Formula C24H24N4O3. MW 416.48. Synthetic Photo-stable fluorescent probe for the detection of nitric oxide NO.