Learn More
Enzo Life Sciences DAR-4M (1mg)
SDP
Supplier:  Enzo Life Sciences ALX620067M001
Encompass_Preferred
Fluorescent probe (Ex: ~560nm; Em: 575nm) for the detection of nitric oxide (NO). It can be used to detect NO directly in biopsy tissues, cultured cells, and tissue sections. Stable in a pH range of 4-12. Low decrease in fluorescence intensity over time. Detection limit 7nM. Not for sale in Japan. Alternative name: Diaminorhodamine-4M. Formulation: Dissolved in 0.47ml DMSO. Purity: ≥98% (HPLC). Appearance: Reddish-violet liquid. Long Term Storage: -20°C.
Catalog No. 50-200-9601