Learn More
Enzo Life Sciences DAR-4M AM (cell permeable) (1mg)
SDP
Catalog No. 502009602
Encompass_Preferred
Cell permeable analog of DAR-4M (ALX-620-067). Can be used to detect nitric oxide (NO) directly in cell specimens prepared from various sources, such as the brain (Ex: ~560nm; Em: 575nm). Stable in a pH range of 4-12. Low decrease in fluorescence intensity over time. Detection limit 7nM. Not for sale in Japan. Alternative name: Diaminorhodamine-4M AM. Formulation: Dissolved in 0.32ml DMSO. Purity: ≥98% (HPLC). Long Term Storage: -20°C.
Catalog No. 50-200-9602 Supplier Enzo Life Sciences Supplier No. ALX620069M001
Cell permeable analog of DAR-4M (ALX-620-067). Can be used to detect nitric oxide (NO) directly in cell specimens prepared from various sources, such as the brain (Ex: ~560nm; Em: 575nm). Stable in a pH range of 4-12. Low decrease in fluorescence intensity over time. Detection limit 7nM. Not for sale in Japan. Alternative name: Diaminorhodamine-4M AM. Formulation: Dissolved in 0.32ml DMSO. Purity: ≥98% (HPLC). Long Term Storage: -20°C.