Learn More
AdipoGen DAR-4MT Solution
SDP
Catalog No. 502561058
Encompass_Preferred
Chemical. CAS 339527-82-1. Formula C25H23N5O3. MW 441.49. Synthetic Reference compound for the fluorescent nitric oxide NO probe DAR-4M.
Catalog No. 50-256-1058 Supplier AdipoGen Supplier No. CDXD0520M001
Chemical. CAS 339527-82-1. Formula C25H23N5O3. MW 441.49. Synthetic Reference compound for the fluorescent nitric oxide NO probe DAR-4M.