Learn More
AdipoGen DAR-4T Solution
SDP
Supplier:  AdipoGen CDXD0575M001
Encompass_Preferred
Chemical. Formula C24H21N5O3. MW 427.46. Synthetic. Reference compound for the fluorescent nitric oxide NO probe DAR-4.
Catalog No. 50-256-1053