Learn More
AdipoGen DAR-4T
SDP
Catalog No. 502561149
Encompass_Preferred
Chemical. Formula C24H21N5O3. MW 427.46. Synthetic Reference compound for the fluorescent nitric oxide NO probe DAR-4.
Catalog No. 50-256-1149 Supplier AdipoGen Supplier No. CDXD0275M001
Chemical. Formula C24H21N5O3. MW 427.46. Synthetic Reference compound for the fluorescent nitric oxide NO probe DAR-4.