Learn More
AdipoGen DAR-M
SDP
Supplier:  AdipoGen CDXD0121M005
Encompass_Preferred
Chemical. CAS 261351-44-4. Formula C29H34N4O3. MW 486.62. Synthetic Photo-stable fluorescent probe Ex ~565nm Em 580nm for the detection of nitric oxide NO.
Catalog No. 50-256-1229