Learn More
AdipoGen DAR-MT Solution
SDP
Catalog No. 502561054
Encompass_Preferred
Chemical. CAS 261351-47-7. Formula C29H31N5O3. MW 497.6. Synthetic. Reference compound for the fluorescent nitric oxide NO probe DAR-M.
Catalog No. 50-256-1054 Supplier AdipoGen Supplier No. CDXD0574M001
Chemical. CAS 261351-47-7. Formula C29H31N5O3. MW 497.6. Synthetic. Reference compound for the fluorescent nitric oxide NO probe DAR-M.