Learn More
Selleck Chemical LLC Darapladib
SDP
Catalog No. 5000004243
Encompass_Preferred
Darapladib is a reversible lipoprotein-associated phospholipase A2 (Lp-PLA2) inhibitor with IC50 of 0 25 nM Phase 3
Catalog No. 50-000-04243 Supplier Selleck Chemical LLC Supplier No. S752025MG
Darapladib is a reversible lipoprotein-associated phospholipase A2 (Lp-PLA2) inhibitor with IC50 of 0 25 nM Phase 3