Learn More
Medchemexpress LLC HY-P9915 5mg , Daratumumab CAS:945721-28-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99155MG
Encompass_Preferred
HY-P9915 5mg Daratumumab CAS: 945721-28-8 Daratumumab (Anti-Human CD38) is the first-in-class human-specific anti-CD38 monoclonal antibody (IgG1). Daratumumab has anti-multiple myeloma (MM) effect. Daratumumab impairs MM cell adhesion, which results in an increased sensitivity of MM to proteasome inhibition. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9583