Learn More
Medchemexpress LLC Darbufelone mesylate | 139340-56-0 | 99.7% | 428.57 | C19H28N2O5S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY101438A25MG
Encompass_Preferred
Darbufelone mesylate is a research-grade small molecule inhibitor that suppresses cellular production of PGF2α and LTB4. It potently inhibits PGHS-2 (IC50 = 0.19 μM) with substantially lower activity against PGHS-1 (IC50 = 20 μM), and is provided as the methanesulfonate salt for assay and cell-based studies.
  • Potent PGHS-2 inhibition (IC50 = 0.19 μM).
  • Marked selectivity over PGHS-1 (IC50 = 20 μM).
  • High purity, suitable for analytical and biological use (≈99.7%).
  • Provided as the mesylate salt for improved handling and stability.
  • Characterized molecular weight 428.57 and formula C19H28N2O5S2.
  • Intended for research use in biochemical and cell-based assays.
Catalog No. 50-002-47053