Learn More
Medchemexpress LLC Darbufelone mesylate | 139340-56-0 | MFCD00906463 | 99.7% | 428.57 | C19H28N2O5S2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY101438A50MG
Encompass_Preferred
Darbufelone mesylate is a research-grade small molecule that inhibits cellular production of prostaglandin F2α and leukotriene B4, with preferential activity against PGHS-2. It is supplied as a solid, high-purity salt suitable for in vitro pharmacology and assay development.
  • Dual inhibitor of PGF2α and LTB4 production.
  • Potent PGHS-2 inhibition (IC50 = 0.19 μM).
  • High-purity solid suitable for research applications.
  • Available in small pack sizes for assay development.
  • Datasheet, COA, and SDS available for quality and safety information.
Catalog No. 50-002-47054