Learn More
CPC Scientific H-Tyr-DArg-Phe-Gly-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OPIO006A
Encompass
Peptide that produces significant antinociception in morphine-tolerant mice. It is also the most potent of the (DArg2)-dermorphin fragments and more stable to enzymatic cleavage.SEQUENCE: H-Tyr-DArg-Phe-Gly-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Y-DArg-FG-NH2MOLECULAR FORMULA: C26H36N8O5MOLECULAR WEIGHT: 540.62STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [100304-60-7]SYNONYMS: (D-Arg)-Dermorphin (1-4) amideRESEARCH AREA: PainREFERENCES:K. Suzuki et al., Peptide Chemistry, 1985, 203 (1986)SKU(s): OPIO-006A
Catalog No. 50-210-9977