Learn More
TARGETMOL CHEMICALS INC DARGLITAZONE 10MG
SDP
Catalog No. 502580932
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Darglitazone (CP-86325) is a potent selective agonist of PPAR-(gamma) with antidiabetic actions. Darglitazone is effective in controlling blood glucose and lipid metabolism. purity: 99%
Catalog No. 50-258-0932 Supplier TARGETMOL CHEMICALS INC Supplier No. T2270810MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Darglitazone (CP-86325) is a potent selective agonist of PPAR-(gamma) with antidiabetic actions. Darglitazone is effective in controlling blood glucose and lipid metabolism. purity: 99%