Learn More
Medchemexpress LLC Darifenacin | 133099-04-4 | 99.8% | C28H30N2O2 | 5 MG
SDP
Catalog No. 5000432413
Encompass_Preferred
Darifenacin is a selective and orally active M3 muscarinic receptor (M3R) antagonist. It is primarily used in the study of urinary incontinence and symptoms of overactive bladder. Additionally, it has demonstrated potential anti-tumor effects by inhibiting tumor growth in colorectal cancer cells.
  • Selective M3 muscarinic receptor antagonist (pKi of 8.9).
  • Binds >20-fold more specifically to M3R.
  • Inhibits tumor growth in colorectal cancer cells.
  • Used in studies for urinary incontinence and overactive bladder.
Catalog No. 50-004-32413 Supplier Medchemexpress LLC Supplier No. HYA00335MG
Darifenacin is a selective and orally active M3 muscarinic receptor (M3R) antagonist. It is primarily used in the study of urinary incontinence and symptoms of overactive bladder. Additionally, it has demonstrated potential anti-tumor effects by inhibiting tumor growth in colorectal cancer cells.
  • Selective M3 muscarinic receptor antagonist (pKi of 8.9).
  • Binds >20-fold more specifically to M3R.
  • Inhibits tumor growth in colorectal cancer cells.
  • Used in studies for urinary incontinence and overactive bladder.