Learn More
Cayman Chemical Darifenacin-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1189701-43-6, 10 mg
SDP
Catalog No. 502947559
Encompass_Preferred
An internal standard for the quantification of darifenacin by GC- or LC-MS
Catalog No. 50-294-7559 Supplier Cayman Chemical Supplier No. 3370110 MG
An internal standard for the quantification of darifenacin by GC- or LC-MS