Learn More
Cayman Chemical DarIfenacIn-d4 500ug
SDP
Catalog No. 502947947
Encompass_Preferred
An internal standard for the quantification of darifenacin by GC- or LC-MS
Catalog No. 50-294-7947 Supplier Cayman Chemical Supplier No. 33701500 UG
An internal standard for the quantification of darifenacin by GC- or LC-MS