Learn More
Medchemexpress LLC (±)-Darifenacin ((±)-UK-88525) | 133033-93-9 | 99.4% | 426.55 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY22437100MG
Encompass_Preferred
(±)-Darifenacin is the racemate of Darifenacin, which is a selective M3 muscarinic receptor antagonist.
  • Soluble in DMSO at 100 mg/mL (234.44 mM).
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
  • Appears as a solid, white to off-white.
  • Related to neurological, eye or ear disease, cancer, neurodegenerative disease, Parkinson's disease, cancer metabolism and metastasis.
  • Involved in GPCR/G protein, neuronal signaling, mAChR pathways.
Catalog No. 50-004-00542