Description
- For any transmitted light stand (halogen or LED).
- Iris diaphragm allows you to vary the illumination aperture according to the objective numerical aperture.
Specifications
Specifications
| Type | Darkfield Attachment |
| For Use With (Equipment) | Halogen or LED transmitted light stand, SMZ-171 Microscopes |
| Includes | Iris diaphragm |