Learn More
Electron Microscopy Sciences Darkroom Fan, Model Sl/Wc
SDP
Catalog No. 5025829
Encompass_Preferred
With all of the toxic chemicals and fumes which are released during film processing, the need for adequate ventilation in darkroom work areas cannot be over emphasized. Improperly ventilated space can be harmful to the health of workers. Suited to exhaust air through a cinderblock wall or stud partition dry wall. Comes with outside exhaust wall cap and flap and grill to keep out wild life. Airflow is from 600-900 CFM. For up 20' square room.
Catalog No. 50-258-29 Supplier Electron Microscopy Sciences Supplier No. 7485040
With all of the toxic chemicals and fumes which are released during film processing, the need for adequate ventilation in darkroom work areas cannot be over emphasized. Improperly ventilated space can be harmful to the health of workers. Suited to exhaust air through a cinderblock wall or stud partition dry wall. Comes with outside exhaust wall cap and flap and grill to keep out wild life. Airflow is from 600-900 CFM. For up 20' square room.