Learn More
Electron Microscopy Sciences Darkroom Louver, Model L-1212
SDP
Catalog No. 5026979
Encompass_Preferred
Fresh air supply louvers for use with darkroom fans. Made of molded plastic. Use with fan A-1212.
Catalog No. 50-269-79 Supplier Electron Microscopy Sciences Supplier No. 7485120
Fresh air supply louvers for use with darkroom fans. Made of molded plastic. Use with fan A-1212.