Learn More
Electron Microscopy Sciences Darkroom Louver, Model L-1224
SDP
Catalog No. 5026980
Encompass_Preferred
Fresh air supply louvers for use with darkroom fans. Made of molded plastic. Use with fans Pro-Lab SL/PS and Pro-Lab SL/WC.
Catalog No. 50-269-80 Supplier Electron Microscopy Sciences Supplier No. 7485130
Fresh air supply louvers for use with darkroom fans. Made of molded plastic. Use with fans Pro-Lab SL/PS and Pro-Lab SL/WC.