Learn More
Electron Microscopy Sciences Darkroom Thermometer, Model A
SDP
Catalog No. 5025290
Encompass_Preferred
Precision Dial Thermometer: easy to read Fahrenheit and Centigrade scales. Hermetically sealed; water proof; will not fog; made from corrosion resistant stainless steel. Comes with an adjustable positioning clamp. Accuracy to 1/2 degree. With 1 3/4" dial with a 6" stem.
Catalog No. 50-252-90 Supplier Electron Microscopy Sciences Supplier No. 74388
Precision Dial Thermometer: easy to read Fahrenheit and Centigrade scales. Hermetically sealed; water proof; will not fog; made from corrosion resistant stainless steel. Comes with an adjustable positioning clamp. Accuracy to 1/2 degree. With 1 3/4" dial with a 6" stem.