Learn More
Electron Microscopy Sciences Darkroom Timerolt1/19/201, 220 V
SDP
Catalog No. 5027861
Encompass_Preferred
A new improved Model 300 Darkroom Timer shows minutes and seconds in total darkness. The 60-minute time range can be set in minutes for developing and seconds for enlarging control. . Two separate outlet receptacles control an enlarger or printer and safelight by a combination time/focus switch. Each outlet has a 750-watts capacity.
Catalog No. 50-278-61 Supplier Electron Microscopy Sciences Supplier No. 7439522
A new improved Model 300 Darkroom Timer shows minutes and seconds in total darkness. The 60-minute time range can be set in minutes for developing and seconds for enlarging control. . Two separate outlet receptacles control an enlarger or printer and safelight by a combination time/focus switch. Each outlet has a 750-watts capacity.