Learn More
Medchemexpress LLC HY-16985 50mg , Darolutamide CAS:1297538-32-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1698550MG
Encompass_Preferred
HY-16985 50mg Darolutamide CAS: 1297538-32-9 Darolutamide (ODM-201;BAY-1841788) is a potent androgen receptor (AR) antagonist with an IC50 of 26 nM in in vitro assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0990